4-Phenoxybenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: p-Phenoxybenzenecarbaldehyde, 4-Phenoxybenzenecarbaldehyde

P1333-25G P1333-5G
TCP1333-025GEA 391.84 CAD
TCP1333-025G TCP1333-005G
4-Phenoxybenzaldehyde ≥98.0%
4-Phenoxybenzaldehyde

CAS Number: 67-36-7
MDL Number: MFCD00003383
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 185
Flash Point (°C): 112
Freezing point (°C): 25


Other Product Names / Synonyms: 4-Formyldiphenyl Ether

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 185 °C
Melting Pt: 24.5 °C
Density: 1.1522 g/cm³ (20 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003383
CAS Number: 67-36-7
EINECS: 200-650-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d41df5cf919e5d537953a774b383181cc27d92e&vid=b751a9712efee5f3924f295c4e96e3d66445cb02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR