5-Phenyl-1-pentanol ≥98.0%

Supplier: TCI America

P1325-5G P1325-25G
TCP1325-005GEA 252.34 CAD
TCP1325-005G TCP1325-25G
5-Phenyl-1-pentanol ≥98.0%
5-Phenyl-1-pentanol

CAS Number: 10521-91-2
MDL Number: MFCD00002979
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 152
Flash Point (°C): 110
Specific Gravity (20/20): 0.97

Formula: (C₆H₅)CH₂CH₂CH₂CH₂CH₂OH
Boiling Pt: 152 °C
Density: 0.97 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002979
CAS Number: 10521-91-2
EINECS: 234-064-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e66772f72075e819fb45c56ada4b51a06c1ebc57&vid=e66772f72075e819fb45c56ada4b51a06c1ebc57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR