DL-Panthenol ≥98.0%

Supplier: TCI America

Synonyms: Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

P1318-500G P1318-25G
TCP1318-500GEA 285.98 CAD
TCP1318-500G TCP1318-025G
DL-Panthenol ≥98.0%
DL-Panthenol

CAS Number: 16485-10-2
MDL Number: MFCD00002944
Molecular Formula: C9H19NO4
Molecular Weight: 205.25
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 66


Other Product Names / Synonyms: 3-(2,4-Dihydroxy-3,3-dimethylbutyramido)-1-propanol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide

Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
Melting Pt: 66 °C
MDL Number: MFCD00002944
CAS Number: 16485-10-2
EINECS: 240-540-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33ab78eab58d4ea6b992d636288d8ec3bdeeb141&vid=33ab78eab58d4ea6b992d636288d8ec3bdeeb141&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR