(R)-(-)-4-Phenyl-2-oxazolidinone ≥99.0%
Supplier: TCI America
Synonyms:
(R)-4-Phenyloxazolidin-2-one
P1307-25G
P1307-5G
TCP1307-025GEA
79
CAD
TCP1307-025G
TCP1307-005G
(R)-(-)-4-Phenyl-2-oxazolidinone ≥99.0%
(R)-(-)-4-Phenyl-2-oxazolidinone
CAS Number: 90319-52-1
MDL Number: MFCD00192393
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 131
Specific rotation [a]20/D: -72 deg (C=1, EtOAc)
Formula:
C₉H₉NO₂ MW: 163.18 g/mol Melting Pt: 131 °C |
MDL Number:
MFCD00192393 CAS Number: 90319-52-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...