Ethyl-3-phenylpropionate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester

P1304-250G P1304-25G
TCP1304-250GEA 376.04 CAD
TCP1304-250G TCP1304-025G
Ethyl-3-phenylpropionate ≥98.0%
Ethyl 3-phenylpropionate

CAS Number: 2021-28-5
MDL Number: MFCD00009206
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 248
Flash Point (°C): 108
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Ethyl Hydrocinnamate, 3-Phenylpropionic Acid Ethyl Ester

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 248 °C
Density: 1.02 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00009206
CAS Number: 2021-28-5
EINECS: 217-966-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5a0f094458f5c1d2b0dfe7b1efd2c253b303d4b&vid=b5a0f094458f5c1d2b0dfe7b1efd2c253b303d4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR