Cyclohexen-1-ylbenzene ≥95.0%

Supplier: TCI America

P1302-25ML P1302-5ML
TCP1302-025MLEA 218.04 CAD
TCP1302-025ML TCP1302-005ML
Cyclohexen-1-ylbenzene ≥95.0%
Cyclohexen-1-ylbenzene

CAS Number: 771-98-2
MDL Number: MFCD00001542
Molecular Formula: C12H14
Molecular Weight: 158.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 252
Flash Point (°C): 103
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 1-Cyclohexen-1-ylbenzene

Formula: C₁₂H₁₄
Boiling Pt: 252 °C
Melting Pt: 0 °C
Density: 0.99 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00001542
CAS Number: 771-98-2
EINECS: 212-242-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d966d766c789bf4335f8e194ccb34b7733a579c&vid=18b20d2b83fc9bd01b1f1084a9de36a9824ad88b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR