Pentaerythrityl tetrachloride ≥98.0%

Supplier: TCI America

Synonyms: Pentaerythrityl Tetrachloride

P1295-5G P1295-25G
TCP1295-5GEA 124.03 CAD
TCP1295-5G TCP1295-025G
Pentaerythrityl tetrachloride ≥98.0%
Pentaerythrityl tetrachloride

CAS Number: 3228-99-7
MDL Number: MFCD00000942
Molecular Formula: C5H8Cl4
Molecular Weight: 209.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 110
Melting point (°C): 96


Other Product Names / Synonyms: 1,3-Dichloro-2,2-bis(chloromethyl)propane, Tetrakis(chloromethyl)methane

Formula: C₅H₈Cl₄
MW: 209.93 g/mol
Boiling Pt: 110 °C
Melting Pt: 96 °C
Density: 1.675 g/cm³ (20 °C)
MDL Number: MFCD00000942
CAS Number: 3228-99-7
EINECS: 221-763-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c3d56beb4d8e2eff12aa42640cd4986a285ddfd&vid=e0076aa801cd8f0883ff15da71da0e1c606c04c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR