(S)-(+)-β-Aminophenethyl alcohol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P1294-25G P1294-5G
TCP1294-25GEA 106.07 CAD
TCP1294-25G TCP1294-005G
(S)-(+)-β-Aminophenethyl alcohol ≥97.0% (by GC, titration analysis)
(S)-(+)-β-Aminophenethyl alcohol

CAS Number: 20989-17-7
MDL Number: MFCD00064404
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 74
Specific rotation [a]20/D: 30 deg (C=1, EtOH)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-2-Amino-2-phenylethanol

Formula: C₈H₁₁NO
Melting Pt: 74 °C
MDL Number: MFCD00064404
CAS Number: 20989-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d3f581023bf1abc8ae03e296fd656c777ab6369&vid=0d3f581023bf1abc8ae03e296fd656c777ab6369&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR