4-Phenyl-1-butanol ≥95.0%

Supplier: TCI America

Synonyms: 4-Phenylbutan-1-ol

P1275-25ML P1275-5ML
TCP1275-025MLEA 209.98 CAD
TCP1275-025ML TCP1275-005ML
4-Phenyl-1-butanol ≥95.0%
4-Phenyl-1-butanol

CAS Number: 3360-41-6
MDL Number: MFCD00002971
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 4-Phenylbutyl Alcohol

Formula: (C₆H₅)(CH₂)₃CH₂OH
MW: 150.22 g/mol
Boiling Pt: 140 °C
Density: 0.99 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS Number: 3360-41-6
EINECS: 222-128-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c55f7055592d52a187544d8dc116b723d1f265e7&vid=c55f7055592d52a187544d8dc116b723d1f265e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR