(R)-(-)-Prolinol ≥99.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol

P1274-25G P1274-5G P1274-1G
TCP1274-25GEA 541.94 CAD
TCP1274-25G TCP1274-005G TCP1274-001G
(R)-(-)-Prolinol ≥99.0% (by GC, titration analysis)
(R)-(-)-Prolinol

CAS Number: 68832-13-3
MDL Number: MFCD00064321
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 76
Flash Point (°C): 86
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: -30.5 deg (C=1, Toluene)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-2-(Hydroxymethyl)pyrrolidine, (R)-(-)-2-Pyrrolidinemethanol

Formula: C₅H₁₁NO
MW: 101.1 g/mol
Boiling Pt: 76 °C
Density: 1.04 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00064321
CAS Number: 68832-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=938d9fff00f697d59399c47c90e486b25eddc74c&vid=938d9fff00f697d59399c47c90e486b25eddc74c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR