2-Pyrrolecarbaldehyde ≥98.0%
Supplier: TCI America
P1246-5G
P1246-25G
TCP1246-005GEA
61.62
CAD
TCP1246-005G
TCP1246-025G
2-Pyrrolecarbaldehyde ≥98.0%
2-Pyrrolecarbaldehyde
CAS Number: 1003-29-8
MDL Number: MFCD00005217
Molecular Formula: C5H5NO
Molecular Weight: 95.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 219
Melting point (°C): 45
Flash Point (°C): 107
Other Product Names / Synonyms: 2-Formylpyrrole
Formula:
C₅H₅NO Boiling Pt: 219 °C Melting Pt: 45 °C Flash Pt: 107 °C |
MDL Number:
MFCD00005217 CAS Number: 1003-29-8 EINECS: 213-705-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...