Pentylene di(acetate)
Supplier: TCI America
P1238-500G
P1238-25G
TCP1238-500GEA
328.64
CAD
TCP1238-500G
TCP1238-025G
Pentylene di(acetate)
Pentylene di(acetate)
CAS Number: 6963-44-6
MDL Number: MFCD00014985
Molecular Formula: C9H16O4
Molecular Weight: 188.22
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 108
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: Pentamethylene Diacetate, 1,5-Pentanediol Diacetate
Formula:
C₉H₁₆O₄ Boiling Pt: 108 °C Melting Pt: 2 °C Density: 1.03 (20 °C) |
MDL Number:
MFCD00014985 CAS Number: 6963-44-6 EINECS: 230-161-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...