Pentafluoronitrobenzene ≥97.0%
Supplier: TCI America
|
Warning
|
P1228-5G
P1228-25G
TCP1228-005GEA
173.8
CAD
TCP1228-005G
TCP1228-025G
Pentafluoronitrobenzene ≥97.0%
Pentafluoronitrobenzene
CAS Number: 880-78-4
MDL Number: MFCD00007049
Molecular Formula: C6F5NO2
Molecular Weight: 213.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 160
Flash Point (°C): 91
Specific Gravity (20/20): 1.71
Formula:
C₆F₅NO₂ Boiling Pt: 160 °C Density: 1.71 (20 °C) Flash Pt: 91 °C |
MDL Number:
MFCD00007049 CAS Number: 880-78-4 EINECS: 212-915-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...