Pyridine-2,3-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
P1222-5G
P1222-25G
TCP1222-5GEA
51.19
CAD
TCP1222-5G
TCP1222-025G
Pyridine-2,3-dicarboxylic anhydride ≥95.0% (by titrimetric analysis)
Pyridine-2,3-dicarboxylic anhydride
CAS Number: 699-98-9
MDL Number: MFCD00005915
Molecular Formula: C7H3NO3
Molecular Weight: 149.11
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 138
Other Product Names / Synonyms: Quinolinic Anhydride
Formula:
C₇H₃NO₃ Boiling Pt: 360 °C (1013 hPa) Melting Pt: 138 °C |
MDL Number:
MFCD00005915 CAS Number: 699-98-9 EINECS: 211-834-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...