(R)-(-)-β-Aminophenethyl alcohol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: D-Phenylglycinol, (R)-(-)-2-Phenylglycinol

P1201-5G P1201-25G
TCP1201-005GEA 82.63 CAD
TCP1201-005G TCP1201-025G
(R)-(-)-β-Aminophenethyl alcohol ≥98.0% (by GC, titration analysis)
(R)-(-)-β-Aminophenethyl alcohol

CAS Number: 56613-80-0
MDL Number: MFCD00008062
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 79
Specific rotation [a]20/D: -29.5 deg (C=1, EtOH)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-2-Amino-2-phenylethanol

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00008062
CAS Number: 56613-80-0
EINECS: 260-287-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=597beed6058434d862d94dc9ea64dbc7b50c4939&vid=597beed6058434d862d94dc9ea64dbc7b50c4939&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR