(1-Propyl)triphenylphosphonium bromide ≥98.0%

Supplier: TCI America
Warning

Synonyms: Triphenyl(propyl)phosphonium bromide, Triphenylpropylphosphonium bromide

P1200-500G P1200-25G
TCP1200-500GEA 458.2 CAD
TCP1200-500G TCP1200-025G
(1-Propyl)triphenylphosphonium bromide ≥98.0%
(1-Propyl)triphenylphosphonium bromide

CAS Number: 6228-47-3
MDL Number: MFCD00011843
Molecular Formula: C21H22BrP
Molecular Weight: 385.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 239


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Propyltriphenylphosphonium Bromide

Formula: C₂₁H₂₂BrP
MW: 385.28 g/mol
Melting Pt: 239 °C
Storage Temperature: Ambient
MDL Number: MFCD00011843
CAS Number: 6228-47-3
EINECS: 228-330-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ba9e20dab87d0ea3e20a819c4234575860c538a&vid=7ba9e20dab87d0ea3e20a819c4234575860c538a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR