N-(Bromomethyl)phthalimide ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(Bromomethyl)isoindoline-1,3-dione

P1192-25G P1192-5G
TCP1192-025GEA 121.66 CAD
TCP1192-025G TCP1192-005G
N-(Bromomethyl)phthalimide ≥96.0% (by titrimetric analysis)
N-(Bromomethyl)phthalimide

CAS Number: 5332-26-3
MDL Number: MFCD00005897
Molecular Formula: C9H6BrNO2
Molecular Weight: 240.06
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 151


Other Product Names / Synonyms: N-Phthalimidomethyl Bromide

Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Boiling Pt: 410
Melting Pt: 151 °C
MDL Number: MFCD00005897
CAS Number: 5332-26-3
EINECS: 226-239-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e46f64a6d92595e9d5f63830e7ba15e7d233c7&vid=91e46f64a6d92595e9d5f63830e7ba15e7d233c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR