1,1-Dimethoxy-2-phenylethane ≥97.0%
Supplier: TCI America
Synonyms:
Phenylacetaldehyde dimethyl acetal
P1161-25ML
P1161-500ML
TCP1161-025MLEA
30.56
CAD
TCP1161-025ML
TCP1161-500ML
1,1-Dimethoxy-2-phenylethane ≥97.0%
1,1-Dimethoxy-2-phenylethane
CAS Number: 101-48-4
MDL Number: MFCD00008487
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 108
Flash Point (°C): 90
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: (2,2-Dimethoxyethyl)benzene
Formula:
C₆H₅CH₂CH(OCH₃)₂ Boiling Pt: 108 °C Melting Pt: 62…64 °C Density: 1.01 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00008487 CAS Number: 101-48-4 EINECS: 202-945-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...