Pentaethylene glycol ≥95.0%

Supplier: TCI America

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

P1154-5G P1154-25G
TCP1154-005GEA 139.04 CAD
TCP1154-005G TCP1154-25G
Pentaethylene glycol ≥95.0%
Pentaethylene glycol

CAS Number: 4792-15-8
MDL Number: MFCD00002878
Molecular Formula: C10H22O6
Molecular Weight: 238.28
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 184
Flash Point (°C): 113
Specific Gravity (20/20): 1.13

Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Boiling Pt: 184 °C
Density: 1.13 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ff82f2d09b7d5478d487afd7d38fc06d1e3fde1&vid=9ff82f2d09b7d5478d487afd7d38fc06d1e3fde1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR