(R)-(-)-1,2-Propanediol ≥97.0%

Supplier: TCI America

P1152-5G P1152-25G
TCP1152-005GEA 53.72 CAD
TCP1152-005G TCP1152-25G
(R)-(-)-1,2-Propanediol ≥97.0%
(R)-(-)-1,2-Propanediol

CAS Number: 4254-14-2
MDL Number: MFCD00066248
Molecular Formula: C3H8O2
Molecular Weight: 76.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 103
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: -17 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-1,2-Dihydroxypropane, (R)-(-)-Propylene Glycol

Formula: C₃H₈O₂
Density: 1.04 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00066248
CAS Number: 4254-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19c0e4483d48bd2e629ae701d1c8d62c07bc314f&vid=19c0e4483d48bd2e629ae701d1c8d62c07bc314f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR