(S)-(+)-1-Phenyl-1,2-ethanediol ≥98.0%

Supplier: TCI America

P1151-5G P1151-1G
TCP1151-005GEA 338.91 CAD
TCP1151-005G TCP1151-001G
(S)-(+)-1-Phenyl-1,2-ethanediol ≥98.0%
(S)-(+)-1-Phenyl-1,2-ethanediol

CAS Number: 25779-13-9
MDL Number: MFCD00066256
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 82
Melting point (°C): 68
Specific rotation [a]20/D: 40 deg (C=3, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-Styrene Glycol, (S)-(+)-Phenylethylene Glycol

Formula: C₈H₁₀O₂
Boiling Pt: 82 °C
Melting Pt: 68 °C
MDL Number: MFCD00066256
CAS Number: 25779-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca693d973f51d872010b8dc310195fb425486e10&vid=ca693d973f51d872010b8dc310195fb425486e10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR