(R)-(-)-1-Phenyl-1,2-ethanediol ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-Phenylethane-1,2-diol, (R)-(-)-α,β-Dihydroxyethylbenzene, (R)-(-)-Phenylethane-1,2-diol, (R)-(-)-Phenyl-1,2-ethanediol

P1150-1G P1150-5G
TCP1150-001GEA 48.98 CAD
TCP1150-001G TCP1150-005G
(R)-(-)-1-Phenyl-1,2-ethanediol ≥98.0%
(R)-(-)-1-Phenyl-1,2-ethanediol

CAS Number: 16355-00-3
MDL Number: MFCD00064262
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 82
Melting point (°C): 68
Specific rotation [a]20/D: -40 deg (C=3, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-Styrene Glycol, (R)-(-)-Phenylethylene Glycol

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 82 °C
Melting Pt: 68 °C
MDL Number: MFCD00064262
CAS Number: 16355-00-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa59820f81d0865da62361666544a000594e5b6e&vid=aa59820f81d0865da62361666544a000594e5b6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR