3-Phenyl-2-propyn-1-ol ≥97.0%
Supplier: TCI America
P1147-5G
P1147-25G
TCP1147-005GEA
137.14
CAD
TCP1147-005G
TCP1147-25G
3-Phenyl-2-propyn-1-ol ≥97.0%
3-Phenyl-2-propyn-1-ol
CAS Number: 1504-58-1
MDL Number: MFCD00040914
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 110
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-Hydroxymethyl-2-phenylacetylene, 3-Phenylpropargyl Alcohol, 1-Phenyl-1-propyn-3-ol
Formula:
C₉H₈O Boiling Pt: 145 °C Density: 1.06 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00040914 CAS Number: 1504-58-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...