9,10-Bis(phenylethynyl)anthracene ≥98.0%

Supplier: TCI America

P1137-1G P1137-5G
TCP1137-001GEA 91.64 CAD
TCP1137-001G TCP1137-5G
9,10-Bis(phenylethynyl)anthracene ≥98.0%
9,10-Bis(phenylethynyl)anthracene

CAS Number: 10075-85-1
MDL Number: MFCD00012050
Molecular Formula: C30H18
Molecular Weight: 378.47
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 253

Formula: C₃₀H₁₈
Melting Pt: 253 °C
MDL Number: MFCD00012050
CAS Number: 10075-85-1
EINECS: 233-210-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=042859c24942c73a2a922d2f84f7c23c42f0ed61&vid=042859c24942c73a2a922d2f84f7c23c42f0ed61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR