(S)-(+)-1,2-Propanediol ≥98.0%

Supplier: TCI America

P1129-5G P1129-25G
TCP1129-005GEA 72.68 CAD
TCP1129-005G TCP1129-25G
(S)-(+)-1,2-Propanediol ≥98.0%
(S)-(+)-1,2-Propanediol

CAS Number: 4254-15-3
MDL Number: MFCD00004539
Molecular Formula: C3H8O2
Molecular Weight: 76.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Flash Point (°C): 107
Specific Gravity (20/20): 1.04
Specific rotation [a]20/D: 16.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-1,2-Dihydroxypropane, (S)-(+)-Propylene Glycol

Formula: C₃H₈O₂
Boiling Pt: 92 °C
Density: 1.04 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004539
CAS Number: 4254-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=556849289fcb70cdc48bade01e4c53ff0a278a8d&vid=556849289fcb70cdc48bade01e4c53ff0a278a8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR