1-Phenylpyrrole ≥98.0%

Supplier: TCI America

P1122-25G P1122-5G
TCP1122-25GEA 448.72 CAD
TCP1122-25G 76220-206
1-Phenylpyrrole ≥98.0%
1-Phenylpyrrole

CAS Number: 635-90-5
MDL Number: MFCD00005343
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 131
Melting point (°C): 59

Formula: C₁₀H₉N
Boiling Pt: 131 °C
Melting Pt: 59 °C
MDL Number: MFCD00005343
CAS Number: 635-90-5
EINECS: 211-242-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09eb4b2a696acd6326c670a2fd56e31046d7a73d&vid=09eb4b2a696acd6326c670a2fd56e31046d7a73d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR