7-Azaindole ≥98.0%
Supplier: TCI America
Synonyms:
1H-Pyrrolo[2,3-b]pyridine
P1117-25G
P1117-5G
TCP1117-25GEA
202.57
CAD
TCP1117-25G
TCP1117-005G
7-Azaindole ≥98.0%
7-Azaindole
CAS Number: 271-63-6
MDL Number: MFCD00005606
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 270
Melting point (°C): 106
Other Product Names / Synonyms: 7-Azaindole
Formula:
C₇H₆N₂ Boiling Pt: 270 °C Melting Pt: 106 °C Density: 1.301 g/cm³ (20 °C) |
MDL Number:
MFCD00005606 CAS Number: 271-63-6 EINECS: 205-981-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...