6-Phenylcaproic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1108-25G P1108-5G
TCP1108-25GEA 1004.88 CAD
TCP1108-25G TCP1108-005G
6-Phenylcaproic acid ≥98.0% (by GC, titration analysis)
6-Phenylcaproic acid

CAS Number: 5581-75-9
MDL Number: MFCD00014381
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 113
Freezing point (°C): 19
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C

Formula: C₁₂H₁₆O₂
Boiling Pt: 201…202 °C (24 mmHg)
Melting Pt: 23 °C
Density: 1.03 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00014381
CAS Number: 5581-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01b46dfcb316d6e5d292a3076b630a80da093a6c&vid=01b46dfcb316d6e5d292a3076b630a80da093a6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR