N-Phenylethylenediamine ≥98.0%

Supplier: TCI America
Danger

P1101-25G P1101-10G
TCP1101-25GEA 660.44 CAD
TCP1101-25G TCP1101-010G
N-Phenylethylenediamine ≥98.0%
N-Phenylethylenediamine

CAS Number: 1664-40-0
MDL Number: MFCD00008162
Molecular Formula: C8H12N2
Molecular Weight: 136.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 263
Flash Point (°C): 110
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: N-(2-Aminoethyl)aniline

Formula: C₆H₅NHCH₂CH₂NH₂
Boiling Pt: 263 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008162
CAS Number: 1664-40-0
EINECS: 216-773-4
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef7d433395dc27fa9b1e326b00a7f7e13055296c&vid=ef7d433395dc27fa9b1e326b00a7f7e13055296c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR