(±)-1-Phenyl-1-pentanol ≥98.0%

Supplier: TCI America

P1086-5ML P1086-25ML
TCP1086-5MLEA 159.58 CAD
TCP1086-5ML TCP1086-025ML
(±)-1-Phenyl-1-pentanol ≥98.0%
(±)-1-Phenyl-1-pentanol

CAS Number: 583-03-9
MDL Number: MFCD00021935
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 137
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: alpha-Butylbenzyl Alcohol

Formula: C₆H₅CH(OH)(CH₂)₃CH₃
Boiling Pt: 137 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00021935
CAS Number: 583-03-9
EINECS: 209-493-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1affababfaeccf9be8a6641297c933131048efe1&vid=1affababfaeccf9be8a6641297c933131048efe1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR