(2-Pyridyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1075-5G P1075-25G
TCP1075-005GEA 54.51 CAD
TCP1075-005G TCP1075-025G
(2-Pyridyl)acetic acid ethyl ester ≥98.0% (by GC, titration analysis)
(2-Pyridyl)acetic acid ethyl ester

CAS Number: 2739-98-2
MDL Number: MFCD00006359
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 122
Flash Point (°C): 110
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Ethyl 2-Pyridineacetate, 2-Pyridylacetic Acid Ethyl Ester, 2-Pyridineacetic Acid Ethyl Ester

Formula: C₉H₁₁NO₂
Boiling Pt: 122 °C
Melting Pt: 86…90 °C
Density: 1.09 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00006359
CAS Number: 2739-98-2
EINECS: 220-365-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR