1,3-Phenylenediacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1064-5G P1064-25G
TCP1064-005GEA 107.44 CAD
TCP1064-005G TCP1064-025G
1,3-Phenylenediacetic acid ≥98.0% (by titrimetric analysis)
1,3-Phenylenediacetic acid

CAS Number: 19806-17-8
MDL Number: MFCD00004341
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 177

Formula: C₁₀H₁₀O₄
Melting Pt: 177 °C
MDL Number: MFCD00004341
CAS Number: 19806-17-8
EINECS: 243-332-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f0998780b11b6205f8c14423bd7dacb96848723&vid=0f0998780b11b6205f8c14423bd7dacb96848723&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR