3-Phenylpyridine ≥94.0%

Supplier: TCI America
Warning

P1040-5G P1040-25G
TCP1040-005GEA 130.84 CAD
TCP1040-005G TCP1040-025G
3-Phenylpyridine ≥94.0%
3-Phenylpyridine

CAS Number: 1008-88-4
MDL Number: MFCD00006380
Molecular Formula: C11H9N
Molecular Weight: 155.20
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 274
Flash Point (°C): 110
Specific Gravity (20/20): 1.09

Formula: C₁₁H₉N
Boiling Pt: 274 °C
Melting Pt: 76…77 °C
Density: 1.09 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006380
CAS Number: 1008-88-4
EINECS: 213-762-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c90e12147a7d573cfc04803e514b66a3c3093078&vid=c90e12147a7d573cfc04803e514b66a3c3093078&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR