2-Phenylpyridine ≥98.0%

Supplier: TCI America
Warning

P1039-5G P1039-25G
TCP1039-005GEA 66.36 CAD
TCP1039-005G TCP1039-25G
2-Phenylpyridine ≥98.0%
2-Phenylpyridine

CAS Number: 1008-89-5
MDL Number: MFCD00006280
Molecular Formula: C11H9N
Molecular Weight: 155.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 270
Flash Point (°C): 110
Specific Gravity (20/20): 1.09

Formula: C₁₁H₉N
Boiling Pt: 270 °C
Melting Pt: 76…77 °C
Density: 1.09 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5
EINECS: 213-763-1
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0f1d22c7e09ad14183a8387d4bec533f1fcd27b&vid=c0f1d22c7e09ad14183a8387d4bec533f1fcd27b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR