4'-Propylacetophenone ≥95.0%

Supplier: TCI America

P1036-500G P1036-25G
TCP1036-500GEA 846.88 CAD
TCP1036-500G TCP1036-025G
4'-Propylacetophenone ≥95.0%
4'-Propylacetophenone

CAS Number: 2932-65-2
MDL Number: MFCD00041359
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 115
Specific Gravity (20/20): 0.98

Formula: C₁₁H₁₄O
Boiling Pt: 115 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00041359
CAS Number: 2932-65-2
EINECS: 220-903-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5fb2789eaa8842d948b5afadcf27805825f9a88&vid=f5fb2789eaa8842d948b5afadcf27805825f9a88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR