1-(2-Pyridyl)piperazine ≥98.0%
Supplier: TCI America
Synonyms:
2-(1-Piperazinyl)pyridine
P1033-5G
P1033-25G
TCP1033-005GEA
131.14
CAD
TCP1033-005G
TCP1033-025G
1-(2-Pyridyl)piperazine ≥98.0%
1-(2-Pyridyl)piperazine
CAS Number: 34803-66-2
MDL Number: MFCD00006216
Molecular Formula: C9H13N3
Molecular Weight: 163.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 122
Flash Point (°C): 110
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: 2-(1-Piperazinyl)pyridine
Formula:
C₉H₁₃N₃ MW: 163.22 g/mol Boiling Pt: 293 Melting Pt: 85…87 °C Density: 1.12 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00006216 CAS Number: 34803-66-2 EINECS: 252-220-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...