1-Bromo-3-phenoxypropane ≥95.0%

Supplier: TCI America

P1027-250G P1027-25G
TCP1027-250GEA 955.43 CAD
TCP1027-250G TCP1027-025G
1-Bromo-3-phenoxypropane ≥95.0%
1-Bromo-3-phenoxypropane

CAS Number: 588-63-6
MDL Number: MFCD00000256
Molecular Formula: C9H11BrO
Molecular Weight: 215.09
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Flash Point (°C): 96
Freezing point (°C): 10
Specific Gravity (20/20): 1.36


Other Product Names / Synonyms: 3-Bromopropyl Phenyl Ether

Formula: (C₆H₅)OCH₂CH₂CH₂Br
Boiling Pt: 138 °C
Density: 1.36 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00000256
CAS Number: 588-63-6
EINECS: 209-623-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5c8efb071e642b9ed8f2e1780ee2d1c86cae389&vid=e5c8efb071e642b9ed8f2e1780ee2d1c86cae389&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR