Dibenzyl phosphonate ≥95.0%

Supplier: TCI America

Synonyms: Dibenzyl phosphite

P1016-250G P1016-25G
TCP1016-250GEA 727.27 CAD
TCP1016-250G TCP1016-025G
Dibenzyl phosphonate ≥95.0%
Dibenzyl phosphonate

CAS Number: 17176-77-1
MDL Number: MFCD00004774
Molecular Formula: C14H15O3P
Molecular Weight: 262.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Flash Point (°C): 110
Specific Gravity (20/20): 1.18


Product Categories: Synthetic Organic Chemistry, Dialkyl Phosphites (Multi-component Reaction)
Other Product Names / Synonyms: Dibenzyl Phosphonate, Phosphonic Acid Dibenzyl Ester, Phosphorous Acid Dibenzyl Ester

Formula: C₁₄H₁₅O₃P
MW: 262.25 g/mol
Boiling Pt: 165 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004774
CAS Number: 17176-77-1
EINECS: 241-226-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41f5b4eeccd764433b30c2cb8ffdb93a7f9367d8&vid=41f5b4eeccd764433b30c2cb8ffdb93a7f9367d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR