Copper(II) phthalocyanine ≥93.0%

Supplier: TCI America

Synonyms: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15

P1006-25G P1006-500G P1006-100G
TCP1006-025GEA 75.84 CAD
TCP1006-025G TCP1006-500G TCP1006-100G
Copper(II) phthalocyanine ≥93.0%
Copper (II) phthalocyanine

CAS Number: 147-14-8
MDL Number: MFCD00010719
Molecular Formula: C32H16CuN8
Molecular Weight: 576.08
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Color: Deep Blue
Melting point (°C): 480


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Molecular Conductors, Solar Cell Research Reagents, Magnetic Metal Complexes (Magnetic Materials), Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocy
Other Product Names / Synonyms: Phthalocyanine Copper(II)

Formula: C₃₂H₁₆CuN₈
MW: 576.08 g/mol
Melting Pt: 480
MDL Number: MFCD00010719
CAS Number: 147-14-8
EINECS: 205-685-1
Merck Index: 12,02584
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69410913341fe14ddae8a3bcbc1835abc453153&vid=e69410913341fe14ddae8a3bcbc1835abc453153&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR