Copper(II) phthalocyanine ≥90.0%
Supplier: TCI America
Synonyms:
29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15
CAS Number: 147-14-8
MDL Number: MFCD00010719
Molecular Formula: C32H16CuN8
Molecular Weight: 576.08
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: Blue
Melting point (°C): 480
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Molecular Conductors, Solar Cell Research Reagents, Magnetic Metal Complexes (Magnetic Materials), Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Building Blocks for Phthalocy
Other Product Names / Synonyms: Phthalocyanine Copper(II)
Formula:
C₃₂H₁₆CuN₈ MW: 576.08 g/mol Melting Pt: 480 |
MDL Number:
MFCD00010719 CAS Number: 147-14-8 EINECS: 205-685-1 Merck Index: 12,02584 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...