(4-Pyridylthio)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1000-500G P1000-25G
TCP1000-500GEA 409.85 CAD
TCP1000-500G TCP1000-025G
(4-Pyridylthio)acetic acid ≥98.0% (by titrimetric analysis)
(4-Pyridylthio)acetic acid

CAS Number: 10351-19-6
MDL Number: MFCD00006424
Molecular Formula: C7H7NO2S
Molecular Weight: 169.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: S-(4-Pyridyl)thioglycolic Acid

Formula: C₇H₇NO₂S
MDL Number: MFCD00006424
CAS Number: 10351-19-6
EINECS: 233-765-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1e1891d5dbbe496de1922e609dc8ee725c5cef2&vid=f1e1891d5dbbe496de1922e609dc8ee725c5cef2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR