D-(+)-Proline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0994-5G P0994-25G
TCP0994-005GEA 118.97 CAD
TCP0994-005G TCP0994-025G
D-(+)-Proline ≥98.0% (by titrimetric analysis)
D(+)-Proline

CAS Number: 344-25-2
MDL Number: MFCD00064317
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 85 deg (C=4, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-D-Pro-OH

Formula: C₅H₉NO₂
Melting Pt: 223 °C
MDL Number: MFCD00064317
CAS Number: 344-25-2
EINECS: 206-452-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5798067ab14069ccc0025d495b51a8f23978d891&vid=5798067ab14069ccc0025d495b51a8f23978d891&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR