3-Phenoxybenzyl alcohol
Supplier: TCI America
|
Warning
|
P0979-25G
P0979-500G
TCP0979-025GEA
105.86
CAD
TCP0979-025G
TCP0979-500G
3-Phenoxybenzyl alcohol
3-Phenoxybenzyl alcohol
CAS Number: 13826-35-2
MDL Number: MFCD00004636
Molecular Formula: C13H12O2
Molecular Weight: 200.24
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 300
Flash Point (°C): 113
Specific Gravity (20/20): 1.15
Other Product Names / Synonyms: 3-(Hydroxymethyl)diphenyl Ether
Formula:
C₁₃H₁₂O₂ Boiling Pt: 300 °C Density: 1.15 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00004636 CAS Number: 13826-35-2 EINECS: 237-525-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...