4-Phenylbenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
P0961-25G
P0961-100G
TCP0961-025GEA
47.87
CAD
TCP0961-025G
TCP0961-100G
4-Phenylbenzoic acid ≥98.0%
4-Phenylbenzoic acid
CAS Number: 92-92-2
MDL Number: MFCD00002553
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 227
Product Categories: Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 4-Phenylbenzoic Acid
Formula:
C₁₃H₁₀O₂ MW: 198.22 g/mol Boiling Pt: 373 Melting Pt: 220 Density: 1.284 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002553 CAS Number: 92-92-2 EINECS: 202-203-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...