3-Phenoxybenzaldehyde ≥97.0%

Supplier: TCI America

Synonyms: 3-Phenoxybenzenecarbaldehyde, m-Phenoxybenzenecarbaldehyde

P0960-500G P0960-25G
TCP0960-500GEA 1033 CAD
TCP0960-500G TCP0960-025G
3-Phenoxybenzaldehyde ≥97.0%
3-Phenoxybenzaldehyde

CAS Number: 39515-51-0
MDL Number: MFCD00003353
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 184
Flash Point (°C): 110
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 3-Formyldiphenyl Ether

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 184 °C
Melting Pt: 13 °C
Density: 1.15 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003353
CAS Number: 39515-51-0
EINECS: 254-487-1
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ffd828669eb936222351263cd9b48dc8325a2d6&vid=c4f9bc10b3c23ac4e5cde97ba222f7ec46702d9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR