4-Phenoxyphenol ≥99.0%
Supplier: TCI America
P0950-25G
P0950-500G
TCP0950-025GEA
112.18
CAD
TCP0950-025G
TCP0950-500G
4-Phenoxyphenol ≥99.0%
4-Phenoxyphenol
CAS Number: 831-82-3
MDL Number: MFCD00002331
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 167
Melting point (°C): 83
Flash Point (°C): 170
Other Product Names / Synonyms: 4-Hydroxydiphenyl Ether
Formula:
C₁₂H₁₀O₂ Boiling Pt: 167 °C Melting Pt: 83 °C Flash Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002331 CAS Number: 831-82-3 EINECS: 212-611-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...