Pyridinium p-toluenesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P0942-25G P0942-500G P0942-100G
TCP0942-025GEA 27.33 CAD
TCP0942-025G TCP0942-500G TCP0942-100G
Pyridinium p-toluenesulfonate ≥98.0% (by titrimetric analysis)
Pyridinium p-toluenesulphonate

CAS Number: 24057-28-1
MDL Number: MFCD00013108
Molecular Formula: C12H13NO3S
Molecular Weight: 251.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 120


Other Product Names / Synonyms: PPTS

Formula: C₁₂H₁₃NO₃S
Melting Pt: 120 °C
MDL Number: MFCD00013108
CAS Number: 24057-28-1
EINECS: 246-002-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6641d8938b173133dbd0c51aab0a2a56b1e575f&vid=d6641d8938b173133dbd0c51aab0a2a56b1e575f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR