Pyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
P0940-25G
P0940-500G
TCP0940-025GEA
64.26
CAD
TCP0940-025G
TCP0940-500G
Pyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Pyrazine-2-carboxylic acid
CAS Number: 98-97-5
MDL Number: MFCD00006130
Molecular Formula: C5H4N2O2
Molecular Weight: 124.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Other Product Names / Synonyms: Pyrazinoic Acid
Formula:
C₅H₄N₂O₂ Boiling Pt: 313 °C (1013 hPa) |
MDL Number:
MFCD00006130 CAS Number: 98-97-5 EINECS: 202-718-1 Merck Index: 12,08143 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...