2',3',4',5',6'-Pentafluoroacetophenone ≥98.0%

Supplier: TCI America

P0921-5G P0921-25G
TCP0921-005GEA 96.38 CAD
TCP0921-005G TCP0921-25G
2',3',4',5',6'-Pentafluoroacetophenone ≥98.0%
2',3',4',5',6'-Pentafluoroacetophenone

CAS Number: 652-29-9
MDL Number: MFCD00000296
Molecular Formula: C8H3F5O
Molecular Weight: 210.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 131
Flash Point (°C): 66
Specific Gravity (20/20): 1.51

Formula: C₈H₃F₅O
Boiling Pt: 131 °C
Density: 1.51 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00000296
CAS Number: 652-29-9
EINECS: 211-487-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc4fc0248302c0e701e9b1a3c9434947a5f2fa2f&vid=fc4fc0248302c0e701e9b1a3c9434947a5f2fa2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR