2-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P0906-100G P0906-500G P0906-25G
TCP0906-100GEA 442.4 CAD
TCP0906-100G TCP0906-500G TCP0906-025G
2-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)
2-Phenylbenzoic acid

CAS Number: 947-84-2
MDL Number: MFCD00002463
Molecular Formula: C13H10O2
Molecular Weight: 198.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 344
Melting point (°C): 113


Other Product Names / Synonyms: 2-Phenylbenzoic Acid

Formula: C₁₃H₁₀O₂
Boiling Pt: 344 °C
Melting Pt: 113 °C
Density: 1.458 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002463
CAS Number: 947-84-2
EINECS: 213-432-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5b90ef373641d1cedfbd0e770c8f9bfaa82685&vid=885a9e355d7d2ce4d9e5ebe047d053484531ddfe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR